Human Tagged ORF Clone, GPR172B (SLC52A1)
Fournisseur: OriGene
RC225737L4V
ORIGRC225737L4VEA 2230 EUR InStock
ORIGRC225737L4V
Human Tagged ORF Clone, GPR172B (SLC52A1)
Vecteurs, plasmides et librairies
Lenti ORF particles, SLC52A1 (mGFP-tagged) - Human G protein-coupled receptor 172B (GPR172B), transcript variant 1, 200 µl, >10⁷ TU/ml. This clone was engineered to express the complete ORF with an expression tag. Expression varies depending on the nature of the gene.
The molecular sequence of this clone aligns with the gene accession number as a point of reference only. However, individual transcript sequences of the same gene can differ through naturally occurring variations (e.g. polymorphisms), each with its own valid existence.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité