Human Recombinant PPM1B
Fournisseur: OriGene
TP312918
ORIGTP312918EA 1140 EUR InStock
ORIGTP312918
Human Recombinant PPM1B
Protéines et Peptides
The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways.
  • Expression host: HEK293T
  • Buffer: 25 mM Tris-HCl, 100 mM glycine, pH 7,3, 10% glycerol
Stable for 12 months from the date of receipt of the product under proper storage and handling conditions. Avoid repeated freeze-thaw cycles.
This phosphatase has been shown to dephosphorylate cyclin-dependent kinases (CDKs), and thus may be involved in cell cycle control. Overexpression of this phosphatase is reported to cause cell-growth arrest or cell death. Alternative splicing results in multiple transcript variants encoding different isoforms. Additional transcript variants have been described, but currently do not represent full-length sequences.
Attention: For testing in cell culture applications, please filter before use. Note that you may experience some loss of protein during the filtration process.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité