Human ERBB2
Fournisseur: OriGene
TS422909P5
ORIGTS422909P5EA 1540 EUR InStock
ORIGTS422909P5
Human ERBB2
Protéines et Peptides
HEK293T cells were transiently transfected with TrueORF cDNA plasmid. Transfected cells were cultured for 48 hrs. After harvesting, the cultured cells were fixed in formalin and dehydrated before embedding in paraffin. 5 µm sections of the FFPE cell pellet blocks are cut and mounted on positively charged SuperFrost slides.
  • Expression host: HEK293T
Stability: Slides are guaranteed for a year from the date of receipt if proper storage instructions were followed
Attention: This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité