Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Orion™ Lab Star PH111 Bench pH Meters

Fournisseur: Thermo Fisher Scientific
Orion™
662-2471EA 715 EUR InStock
662-2471 662-2470 662-2474 662-2473 662-2472
Orion™ Lab Star PH111 Bench pH Meters
Compteurs électrochimiques pH-mètres et multi-paramètres
Measure pH or ORP simultaneously with mV and temperature using the Orion Lab Star PH111 pH/mV meter. Featuring a large, backlit display for easy viewability and small footprint to save bench space, this pH meter is ideal for simplified, budget-friendly lab testing. Use the onscreen stabilizing and ready indicators to help identify when pH meter and ORP meter readings are complete. View active pH, ORP and temperature calibration logs with date and time stamp, record key measurements using the 500-point data log with date and time stamp, and output all logs to a printer or computer.

  • Budget-friendly pH meter for simplified pH and ORP testing
  • Display shows pH, mV and temperature simultaneously for accurate data recording
  • Large 5 inch backlit display makes viewing and recording measurements easy
  • Small meter footprint saves valuable bench space in your lab
  • Onscreen stabilising and ready indicators help identify when readings are complete
  • pH electrode condition icon helps ensure reliable electrode performance
  • Date and time data stamped data and calibration logs support GLP compliance
  • Use 500-point data log with date and time stamp to record key measurements
  • Output data and calibration logs to a printer or computer for long-term data storage
  • Three year meter warranty
  • Backlight option is available

Measure −2 to +18 pH, −2000 to +2000 mV and −5 to +105 °C or 23 to 221 °F to ensure comprehensive measurement capabilities. Perform one to five point pH calibrations, one point ORP calibrations and one point temperature adjustment calibrations for proficient SOP compliance. View active pH, ORP and temperature calibration logs with date and time stamp to ensure accurate, reproducible measurements.

It is the pH meter and stand option, which allows you to purchase your pH electrode separately. The standard pH meter kit option, for common pH samples such as drinking water, tap water, buffers (non-TRIS) and general purpose student use.

It is the easy-to-clean bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included refillable pH electrode is easy-to-clean and restore. The low-maintenance bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included gel-filled pH electrode is low-maintenance and easy to operate. It is the difficult sample pH meter kit option, for pH samples that are thick, viscous or semi-solid including foods, beverages, wastewaters, emulsions, soils, slurries, suspended solids or sludges.

AC Adapter: Universal 100 to 240 V 50/60 Hz power supply with twist-and-lock meter connection and interchangeable wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China outlets
Data Management Details: Transfer single or all data points to printer or computer
Data Points: 500 with Date and Time Stamp
Measurement Modes: pH with mV and temperature or ORP with mV and temperature
Log Function Type: Automatic with Auto-Read and Timed Read Modes, Manual with Continuous Read Mode
Range (ORP): ±2000 mV with ±250 mV offset
Temperature Selection: Manual or automatic with ATC temperature probe

Certifications: CE, TUV 3-1, FCC class A.

Information de livraison: Includes: 9157BNMD refillable epoxy-body pH/ATC electrode 916099 pH 4/7/10 buffer, storage and cleaning solution 60 ml kit meter with electrode stand, PC cable, universal power adapter.

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

ACCESSOIRES

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité