Conductivity meter, bench, Orion™ Star™ A212
Fournisseur: Thermo Orion
STARA2126 STARA2120 STARA2125
ORIOSTARA2126EA 1920 EUR InStock
ORIOSTARA2126 663-0212 663-0213
Conductivity meter, bench, Orion™ Star™ A212
Electrochemistry Meters Conductimètres
The Orion™ Star™ A112 conductivity benchtop meter is ideal for a wide range of applications and advanced conductivity analysis in the lab. It reliably measures conductivity, total dissolved solids, salinity, resistivity and temperature values.
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Meter-controlled stirrer probe can be used to uniformly mix solutions
  • Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
  • Calibration log stores ten most recent calibrations
  • Non volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited.
Perform up to a five point conductivity calibration with calibration editing option that allows individual points to be fixed without a full recalibration. Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate.
Use linear, non linear, EP (USP) or off conductivity temperature compensation with 5, 10, 15, 20 or 25 °C reference temperatures.
AC adapter : Universal 50 to 60 Hz, 100 to 240 VAC power adapter (Cat. No. 1010003)
Provides three years meter warranty.
Informations de commande: For compatible buffer solutions, see Orion™ item 11007.
Information de livraison: Supplied with electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable, and meter test certificate.
Kit 1 additionally includes 013005MD DuraProbe 4-cell (K=0,475) epoxy-body conductivity sensor and Orion 1413 µS conductivity standard (5× 60 ml).
Kit 2 additionally includes 013016MD Pure water 2-cell (K=0,1) conductivity sensor with detachable flow cell and Orion 100 µS conductivity standard (5× 60 ml).
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité