pH / LogR measurement module for Orion™ Versa Star Pro™ meters
Fournisseur: Thermo Orion
VSTAR-LR
ORIOVSTAR-LREA 1090 EUR InStock
ORIOVSTAR-LR
pH / LogR measurement module for Orion™ Versa Star Pro™ meters
Electrochemistry Meters pH-mètres et multi-paramètres
This measurement module can be used to customise the measurement capabilities of the Orion™ Versa Star Pro™ meters. It adds pH, mV, ORP and LogR temperature measurement capabilities to the meter. The module offers LogR temperature technology that uses the glass-bulb of most pH electrodes to measure both pH and temperature at the same location – ideal for small sample volumes and samples that are difficult to clean.
  • Each Orion™ Versa Star Pro meter has four channels that can be customised with interchangeable measurement modules
  • Measurement modules are interchangeable
  • Up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Customise measurements to show calibration history and pH electrode condition or simplify to show an extra large pH reading
Range (pH): −2,000 to +20,000
Range (mV): ±2000,0
Range (Temperature): −5 to +105 °C
3-year replacement warranty.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité