Tick tweezers
Fournisseur: OUTILS RUBIS SA
TICK
OUTITICKEA 26.8 EUR InStock
OUTITICK
Tick tweezers
Pinces
Stainless steel, anti-magnetic. These acid-proof tweezers are ideal for removing ticks. Thanks to the straight, very fine tips that are also angled to the inside, ticks can easily be removed.
  • Also perfect for the removal of thorns and splinters
  • Anti-magnetic and rust-proof surgical steel
  • Made completely by hand - and tested individually under a magnifying glass
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité