Blood culture system, Signal®
Fournisseur: THERMO FISHER DIAGNOSTICS
BC0100M BC0104A
OXOIBC0100MEA 279 EUR InStock
OXOIBC0100M OXOIBC0104A
Blood culture system, Signal®
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons de sang
Blood samples are collected from patients, using strict aseptic technique and sterile equipment. The samples are inoculated into the blood culture bottles and mixed with the medium.
  • Well-proven
  • Reliable
The formulation of the medium encourages the growth of aerobic, anaerobic and micro-aerophilic organisms. The medium is also designed to create pressure in the sealed bottle when organisms are growing. The detection of positive pressure is by means of a growth indicator device which is connected to the bottle after the blood sample is added.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité