Culti-Loops™ Proteus mirabilis 511-116
Fournisseur: THERMO FISHER DIAGNOSTICS GMBH
R4608088
OXOIR4608088EA 0 EUR InStock
OXOIR4608088
Culti-Loops™ Proteus mirabilis 511-116
Tests microbiologiques Tests sur les micro-organismes
Confidently carry out testing of media, reagents, and identification and antimicrobial susceptibility kits and evaluate the bacteriological procedures with Culti-Loops™ ready-to-use QC organisms. Proteus mirabilis 511-116 strain produces extended-spectrum β-lactamase (ESBL). This strain is recommended to be used as a positive control strain with Spectra™ ESBL Medium following the HAI surveillance programs.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Conditionnement: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Attention: For in vitro diagnostic use.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité