Senescence kit
Fournisseur: OZ BIOSCIENCES
GXS0003
OZBIGXS0003EA 199.16 EUR InStock
OZBIGXS0003
Senescence kit
Tests Tests cellulaires
Senescence kit allows visualisation of aging cells and is ideal when cells in culture show abnormalities typical of the Hayflick limit after a long period of normal growth. The cells vary in size and shape, the cytoplasm begins to be granular with many cell inclusions and debris formed in the medium.
  • Simple and rapid
  • Ready to use
  • Economical
Senescence goes with decreasing of proliferation potential, telomere shortening and impairment of cell function. During senescence in mammalian cells, an endogenous lysosomal β-galactosidase is over-expressed and is accumulated within the aging cells. The presence of this β-Gal activity is a marker of aging cell population in vitro.
Store kit at +4 °C.
Information de livraison: Kit includes Fixing Buffer (125 ml), 10X PBS (75 ml), Staining Buffer (125 ml) and 25X Stock Solution of SA X-Gal (4×1 ml).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité