Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD9 Mouse Monoclonal Antibody (ExoBrite™ 650/665)

Fournisseur: Biotium
P003-650-125
P003-650-125EA 168 EUR InStock
P003-650-125
Anti-CD9 Mouse Monoclonal Antibody (ExoBrite™ 650/665)
Anticorps
ExoBrite™ IgG1 isotype control flow antibody has been found to not react with any target in human cells and have the same isotype as the ExoBrite™ Flow Antibodies against tetraspanin proteins CD9. The antibody is available with the same conjugates as ExoBrite™ Flow Antibodies for the Pacific Blue™, FITC, PE, and APC channels.

  • Non-reactive IgG1 mouse antibody
  • Serves as a isotype negative control for ExoBrite™ flow antibodies

Extracellular vesicles (EVs), including exosomes, are lipid-bound vesicles that are released from cells. EVs display specific surface proteins and can carry nucleic acids and other cargo, allowing them to transfer biological information between cells in different parts of the body. Therefore EVs are increasingly studied for their potential use in drug delivery and medical diagnostic applications. The most common proteins used as EV markers are CD9, members of the tetraspanin family. Tetraspanins are plasma membrane proteins with many proposed functions, including activation and sorting of other membrane proteins. They are also thought to play a role in the targeting of proteins to multivesicular bodies (MVBs) and exosomes. These tetraspanins are broadly expressed on many cell types and can therefore be detected on many types of EVs, but their expression levels vary depending on the cell type of origin.

Shelf life: guaranteed for at least 24 months from date of receipt when stored as recommended.

Attention: For research use only.

Type: Control
Antigen: CD9
Clonality: Monoclonal
Clone:
Conjugation: ExoBrite™ 650/665
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Baboon, Bovine, Cynomolgus monkey, Dog, Horse, Human, Non-human primates, Rabbit, Sheep

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité