Antibody labelling kits, CyDye™
Fournisseur: Cytiva
PA35000
PA35000EA 581 EUR InStock
PA35000
Antibody labelling kits, CyDye™
Systèmes de liaison et de marquage des protéines
CyDye Fluorescent Dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
  • Convenient method for labeling antibodies with CyDye Fluorescent Dyes
  • Conjugations are carried out under mild conditions and are complete within 1 hour
  • High purity dyes are packaged in premeasured amounts and foil-sealed to ensure consistent labeling
  • Antibody kits contain all reagents required for 2 × 1 mg reactions
Dyes are packaged in premeasured amounts and foil-sealed to ensure consistent labelings. CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
Informations de commande: Components include two foil packs each containing dried dye to label 1mg of protein; Coupling buffer (2 vials) [1M sodium carbonate buffer, pH 9.3]; Gel filtration column for the separation of labelled protein from non-conjugated dye; Elution buffer (50 ml) for the elution of labelled protein from the column (Phosphate buffered saline, pH 7.2, containing 0.1% sodium azide); safety data sheet; sample collection tubes (4) and transfer pipettes (6).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité