Biopharm pressure sensors, SciLog®
Fournisseur: PARKER HANNIFIN
080-699PSX-5
PARK080-699PSX-5EA 1010 EUR InStock
PARK080-699PSX-5
Biopharm pressure sensors, SciLog®
Fluid Sensors
SciLog® range of pre-calibrated, single-use sensors, can measure and document pressure in a range of filtration, separation and purification applications.
  • Trans-membrane pressure (TMP) control
  • Suitable for single-use GMP applications
  • The sensors are validated for exposure to gamma irradiation, autoclave and sodium hydroxide sterilization
Its applications are growth media preparation, bioreactor maintenance, cell culture harvest, buffer preparation, product filtration, tangential flow filtration, virus control and bulk drug dispense.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité