Human plasma, human immunodeficiency virus (HIV)
Fournisseur: Charles River Laboratories Cell Solutions
PB006C-1-HIV
PB006C-1-HIVEA 0 EUR InStock
PB006C-1-HIV
Human plasma, human immunodeficiency virus (HIV)
Plasma
Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of human immunodeficiency virus (HIV). Collected from IRB-consented donors diagnosed with HIV.
There is no complete medical remedy for HIV or AIDS, however it can be possible to maintain the level of HIV in the body low with proper treatment. This samples have been used by researchers to make biomarkers for diagnostic development, preclinacal research validation and for analysis of drug toxicity and efficacy.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
Attention: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.
Collapse All
Désignation Cdt Numéro de catalogue VWR Quantité
Plasma, human immunodeficiency virus (HIV), cryopreserved 2 mL PB006C-1-HIV
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité