Human serum, colorectal cancer (CRC)
Fournisseur: Charles River Laboratories Cell Solutions
PB007C-1-CRC
PB007C-1-CRCEA 0 EUR InStock
PB007C-1-CRC
Human serum, colorectal cancer (CRC)
Sérums
Human serum used for researching the causes, diagnosis, supportive care and treatment of colorectal cancer (CRC). Collected from IRB-consented donors diagnosed with CRC.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV nucleic acid, WNV nucleic acid, Trypanasoma cruzi and Syphilis.
Storage: Store at −80 °C.
Attention: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products orMore Product Information
Collapse All
Désignation Format du sérum Cdt Numéro de catalogue VWR Quantité
Serum, colorectal cancer (CRC) Cryopreserved 2 mL PB007C-1-CRC
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité