Human serum, hepatitis C virus (HCV)
Fournisseur: Charles River Laboratories Cell Solutions
PB007C-1-HCV
PB007C-1-HCVEA 0 EUR InStock
PB007C-1-HCV
Human serum, hepatitis C virus (HCV)
Sérums
Human serum used for researching the causes, diagnosis, supportive care and treatment of hepatitis C Virus (HCV). Collected from IRB-consented donors diagnosed with HCV.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV nucleic acid, WNV nucleic acid, Trypanasoma cruzi and Syphilis.
Storage: Store at −80 °C.
Attention: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products orMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité