Human serum, human immunodeficiency virus (HIV)
Fournisseur: Charles River Laboratories Cell Solutions
PB007C-1-HIV
PB007C-1-HIVEA 0 EUR InStock
PB007C-1-HIV
Human serum, human immunodeficiency virus (HIV)
Sérums
Human serum used for researching the causes, diagnosis, supportive care and treatment of human immunodeficiency virus (HIV). Collected from IRB-consented donors diagnosed with HIV.
There is currently no cure for HIV or AIDS, however with proper treatment we are able to keep the level of HIV virus in the body low. This donors samples can be used by researchers to investigate biomarker and to develop the diagnostic methodologies, pre-clinical research validation, and drug toxicity and efficacy.
Storage: Store at −80 °C.
Attention: Intended for research use only and not for directMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité