UHP nitrogen generator, NG5000
Fournisseur: PEAK SCIENTIFIC
10-2320
PEAK10-2320EA 12570 EUR InStock
PEAK10-2320
UHP nitrogen generator, NG5000
Générateurs de gaz Générateurs d'azote
The NG5000 delivers ultra high purity nitrogen at trace detection limits through Pressure Swing Adsorption technology at flows of up to 5 L/min for GC, DSC and DMA applications as well as other laboratory. This generator requires a high quality source of in-house air to supply the unit.
  • Suitable for applications requiring ultra high purity nitrogen
  • Available with or without an external compressor
  • Contains self-regenerating Carbon Molecular Sieve column
  • Gas is supplied on demand so generator works to your schedule
  • 1 year comprehensive on-site warranty
These separation technologies remove oxygen and other impurities in air, delivering hassle-free, ultra high purity nitrogen, on-demand for the laboratory.
Outlet pressure: 5,5 bar
Weight: 80 kg
Certifications: Conforms with: Low Voltage Directive 2014/35/EU; EN 61010-1: 2010 Electrical Equipment for measurement, control and laboratory use.
Electromagnetic Compatibility Directive 2014/30/EU EN 61326-1: 2013 Electrical Equipment for measurement, control and laboratory use.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité