Nitrogen generator, Infinity 1031
Fournisseur: PEAK SCIENTIFIC
13-1270
PEAK13-1270EA 14360 EUR InStock
PEAK13-1270
Nitrogen generator, Infinity 1031
Générateurs de gaz Générateurs d'azote
Membrane technology is at the heart of the Infinity 1031, generating laboratory-grade nitrogen on-site with an existing air supply. With the ability to perform at its maximum 24 hours a day, if required, and minimum maintenance requirements, the Infinity 1031 is a durable, hassle-free solution for Sciex LC-MS instruments.
  • 24/7 operation at optimum performance if required
  • Gas is supplied on demand so generator works to your schedule
  • Few moving parts means little maintenance required and ensures long life of the generator
  • Minimal set-up required
  • Completely silent operation
  • 1 year comprehensive on-site warranty
Noise level: 54 dBA
Inlet pressure: 8,2 to 9,9 bar
Outlet pressure:
7,5 (Nitrogen, dry air)
7,5 bar (dry air)
Certifications: Conforms with: Low Voltage Directive 2014/35/EU
EN 61010-1: 2010 Electrical Equipment for measurement, control and laboratory use.
Electromagnetic Compatibility Directive 2014/30/EU
EN 61326-1: 2013 Electrical Equipment for measurement, control and laboratory use.
ISO8753-1:2010 Class 1.4.1.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité