HTRF 96-Well, White Plate, Low Volume
Fournisseur: PerkinElmer
66PL96005
PERC66PL96005EA 0 EUR InStock
PERC66PL96005
HTRF 96-Well, White Plate, Low Volume
Microplaques Microplaques immunologiques et bandelettes
These plates have been specifically designed as an alternative to 384-well plate small sample series or simply to try HTRF in a convenient environment.
  • HTRF with ELISA plate ease of use and low volume wells for low sample consumption
  • These 96-well low volume plates can be used with all HTRF assays
96-well low volume plates can be used with all HTRF assays, example second messengers, phospho-proteins, cytokines and biomarkers. you can be used in a no-transfer protocol with suspension cells or with a transfer step adherent cells. see plates can be read on all HTRF certified readers, thus making or implementation in your experiments really easy.
Well shape: Round
Well-bottom shape: V-bottom
Attention: For research use only. Not for use in diagnostic procedures.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité