Chromium standard solution, for ICP-MS, Pure Plus grade
Fournisseur: PerkinElmer
N9303736
PERCN9303736EA 106 EUR InStock
PERCN9303736
Chromium standard solution, for ICP-MS, Pure Plus grade
Étalons Étalons cationiques Standards de chrome
Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
  • Analysed by ICP-OES
  • Analysed by classical wet assay
  • 67 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppb level
  • Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Attention: These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité