Shakematic, 1095
Fournisseur: PerkinElmer
PER011090
PERCPER011090EA 0 EUR InStock
PERCPER011090
Shakematic, 1095
Agitateurs Agitateurs rotatifs
The Shakematic 1095 is designed and built specifically for mixing samples for Falling Number analysis.
  • Sample mixing time is reduced to 3 seconds which gives an increase in lab throughput
  • Operator independent results
  • Improved repeatability
  • Increased lab throughput
  • Less manual work
It has enhanced safety features, is CE-labeled and runs significantly quieter and vibration free.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité