AlphaLISA® Streptavidin Acceptor Beads, 250 µg
Fournisseur: PerkinElmer
AL125C
PERKAL125CEA 942 EUR InStock
PERKAL125C
AlphaLISA® Streptavidin Acceptor Beads, 250 µg
Systèmes de liaison et de marquage des protéines
These beads can be used in conjunction with Alpha Donor beads to create AlphaLISA® no-wash assays for: Protein-protein interaction assays, Protein-DNA interaction assays, Protein-RNA interaction assays, protein-small molecule interaction assays, analyte detection assays, biomarker detection assays, antibody detection assays, enzymatic assays, other immunoassays in a typical AlphaLISA assay, 1 mg of acceptor beads is sufficient to run 1,000 to 2,000 wells using a 50 µl reaction volume.
  • Harmonised Code US-3822005090
Features: No-wash steps, no separation steps Ease-of-use: few addition steps, fast assay development Broad range of affinities: detect strong or weak interactions, from pM to mM affinity Distance: measure very large protein or antibody complexes - spanning up to 200 nm or more high avidity: multiple binding sites on each bead enables use of nanomolar concentrations of antibodies or proteins, as well as use of low affinity binders AlphaScreen® and AlphaLISA® are bead-based assay technologies used to study biomolecular interactions in a microplate format. The acronym 'Alpha' stands for amplified luminescent proximity homogeneous assay.
As the name implies, some of the key features of these technologies are that they are non-radioactive, homogeneous proximity assays. Binding of molecules captured on the beads leads to an energy transfer from one bead to the other, ultimately producing a luminescent/fluorescent signal. To understand how a signal is produced, one must begin with an understanding of the beads.
AlphaScreen and AlphaLISA assays require two bead types: Donor beads and Acceptor beads. Each bead type contains a different proprietary mixture of chemicals, which are key elements of the AlphaScreen technology. Donor beads contain a photosensitiser, phthalocyanine, which converts ambient oxygen to an excited and reactive form of O₂, singlet oxygen, upon illumination at 680 nm. Please note that singlet oxygen is not a radical; it is molecular oxygen with a single excited electron. Like other excited molecules, singlet oxygen has a limited lifetime prior to falling back to ground state. Within its 4 µsec half-life, singlet oxygen can diffuse approximately 200 nm in solution.
If an Acceptor bead is within that proximity, energy is transferred from the singlet oxygen to thioxene derivatives within the acceptor bead, subsequently culminating in light production at 520 to 620 nm (AlphaScreen) or at 615 nm (AlphaLISA). In the absence of an acceptor bead, singlet oxygen falls to ground state and no signal is produced. This proximity-dependent chemical energy transfer is the basis for AlphaScreen's homogeneous nature.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité