Pharmaceutical secondary standard, Phenol, certified reference material, Supelco®
Fournisseur: Merck
PHR1047-1G
PHR1047-1GEA 88.4 EUR InStock
PHR1047-1G
Pharmaceutical secondary standard, Phenol, certified reference material, Supelco®
Étalons Étalons organiques
Traceable to USP 1524806.
Phenol may be used as a standard in determining the concentration of phenol present in samples of water using gas chromatography coupled with electron capture detector (GC-ECD).
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=869b4e36b74ad8f3e91532a1c2191238af7c928e&vid=869b4e36b74ad8f3e91532a1c2191238af7c928e&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité