Pharmaceutical secondary standard, Metformin hydrochloride, (CRM), Supelco®
Fournisseur: Merck
PHR1084-500MG
PHR1084-500MGEA 116 EUR InStock
PHR1084-500MG
Pharmaceutical secondary standard, Metformin hydrochloride, (CRM), Supelco®
Étalons Étalons organiques
Metformin is an antidiabetic agent that reduces blood glucose levels and improves insulin sensitivity.
  • Traceable to PhEur M0605000
  • Traceable to USP 1396309
Its metabolic effects, including the inhibition of hepatic gluconeogenesis, are mediated at least in part by activation of the LKB1-AMPK (AMP-activated protein kinase) pathway. Activation of this pathway also appears to be involved in the antiproliferative and proapoptotic actions of metformin in cancer cell lines.
Certifications: This product was designed, produced and verified for accuracy and stability in accordance with ISO/IEC 17025:2005 (AClass Cert AT-1467), ISO GUIDES 34:2009 (AClass Cert AR-1470).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d0b4aa8b0189b9b5bd38d8d9038416fb252a4c9&vid=7d0b4aa8b0189b9b5bd38d8d9038416fb252a4c9&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité