Pharmaceutical secondary standard, cholecalciferol (Vitamin D3), (CRM), Supelco®
Fournisseur: Merck
PHR1237-500MG
PHR1237-500MGEA 117 EUR InStock
PHR1237-500MG
Pharmaceutical secondary standard, cholecalciferol (Vitamin D3), (CRM), Supelco®
Étalons Étalons organiques
These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
  • Traceable to BP 787
  • Traceable to PhEur C2100000
  • Traceable to USP 1131009
Vitamin D acts through a receptor that is a member of the ligand-dependent transcription factor superfamily. Modulates the proliferation and differentiation of both normal and cancer cells. Has antiproliferative and antimetastatic effects on breast, colon, and prostate cancer cells. Activated vitamin D receptors in intestine and bone maintain calcium absorbance and homeostasis.
Certifications: This product was designed, produced and verified for accuracy and stability in accordance with ISO/IEC 17025:2005 (AClass Cert AT-1467), ISO GUIDES 34:2009 (AClass Cert AR-1470).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité