Immunoassay microplates, anti-GST coated, 96-well strip plates, Pierce™
Fournisseur: Thermo Fisher Scientific
15144
PIER15144EA 264 EUR InStock
PIER15144
Immunoassay microplates, anti-GST coated, 96-well strip plates, Pierce™
Microplaques Microplaques immunologiques et bandelettes
Pierce™ Anti-GST coated plates are polystyrene microplates coated with mouse monoclonal anti-GST antibody and pre-blocked for immediate use.
  • Antibody-based - plates are coated with mouse monoclonal anti-GST antibody
  • Sensitive - detects GST fusion proteins down to 7,5 ng/well
  • Flexibility - binds native or denatured forms of GST; an alternative to glutathione-coated plates
  • Ready to use - pre-blocked with SuperBlock Blocking Buffer
  • Compatibility - Thermo Scientific Cell Lysis Reagents and many other popular lysis buffers do not interfere with GST-fusion protein binding
  • Superior performance - improved signal-to-noise ratio relative to competitor's plates
Unlike glutathione coated plates, these anti-GST plates effectively bind both native and denatured forms of GST. In most cases, pre-purification of cell lysates is not necessary before screening and analysis of recombinant GST-tagged protein expression by ELISA using the plates.
Binding property: standard capacity, affinity
Detection method: colorimetric
Surface treatment: antibody
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité