RNA 3' End biotinylation kit, Pierce™
Fournisseur: Thermo Fisher Scientific
20160
PIER20160EA 736 EUR InStock
PIER20160
RNA 3' End biotinylation kit, Pierce™
Systèmes de liaison et de marquage des protéines
Pierce™ RNA 3' End Biotinylation Kit is optimised for labeling the 3' terminal end of RNA probes to facilitate their use as probes or targets in EMSA and other methods for studying protein-RNA interactions.
  • RNA can be labeled in 0,5 to 2 hours with minimal downstream processing
  • Results in minimal disturbance of RNA secondary structure
  • Labels synthetic or in vitro transcribed RNA probes ranging from 22 to 450 nucleotides
  • Incorporates a biotin label which allows detection sensitivity similar to radioactivity when used with chemiluminescent detection systems
Pierce™ RNA 3' End Biotinylation Kit uses T4 RNA ligase to conjugate a single nucleotide analog to the 3' terminus of an RNA strand. The unique biotinylated cytidine (bis)phosphate used in this kit enables T4 RNA ligase to attach a single biotin tag on RNA probes of diverse secondary structure. Labeled RNA is easily precipitated to remove reaction byproducts. The biotin label enables the RNA probe to be immobilized or detected with streptavidin in order to study RNA interactions or to perform non radioactive blotting assays.
Information de livraison: Kit includes T4 RNA ligase reaction buffer, 10X (100 µl); T4 RNA ligase, 20 000 U/ml (40 µl); non-labeled RNA control, 10 µM (100 µl); biotinylated IRE RNA control, 125 nM (35 µl); RNase Inhibitor, 40 U/µl (2×10 µl); nuclease-free water (1,5 ml); DMSO (200 µl); 30% PEG (300 µl); biotinylated cytidine (bis)phosphate, 1 mM (40 µl); and glycogen, 20 mg/ml (20 µl).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité