Magnetic RNA-protein pull-down kit, Pierce™
Fournisseur: Thermo Fisher Scientific
20164
PIER20164EA 932 EUR InStock
PIER20164
Magnetic RNA-protein pull-down kit, Pierce™
Systèmes de liaison et de marquage des protéines
Pierce™ Magnetic RNA-Protein Pull-Down Kit is a streamlined, robust method to enrich protein-RNA interactions using end-labelled RNA as the bait.
  • Captures ribonucleoprotein complexes directly with end-labelled RNA
  • Does not use or require antibodies for pull-down
  • No spin cups or centrifugation necessary for the enrichment of RBP
  • Procedure streamlined for minimal hands-on time (less than 3 hours) after RNA labelling reaction
  • Used in vitro transcribed RNA or synthetic RNA for labelling of various lengths and complexity
  • Low bead background and unrelated RNA or mutated RNA does not significantly enrich specified RBPs
Magnetic RNA-Protein Pull-Down Kit provides reagents to efficiently enrich RNA Binding Proteins (RBPs) using RNA end-labelled with desthiobiotin and streptavidin magnetic beads. This direct enrichment of the protein-RNA interaction provides an alternative to antibody capture of protein-RNA complexes or moieties incorporated into the nucleic acid. The kit is amenable to several downstream applications, including Western blotting and mass spectrometry. It also finds applications in mutational analysis, structure function analaysis and identification of RNA-protein interactions and MS analysis of unknown RNA-protein binding pairs or complexes.
Information de livraison: The kit includes Pierce™ nucleic acid-compatible streptavidin magnetic beads (1 ml), RNA capture buffer (1X) (10 ml), Tris (20 mM, pH 7,5) (5 ml), protein-RNA binding buffer (10X) (1 ml), wash buffer (1X) (10 ml), biotin elution buffer (1,5 ml), HuR monoclonal antibody (mouse) (50 µl), positive RNA control (AR RNA) (250 pmol), negative RNA control (polyA25 RNA) (250 pmol) and Pierce™ RNA 3' end desthiobiotinylation kit.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité