Fluorescent Dye Removal Columns, Pierce™
Fournisseur: Thermo Fisher Scientific
22858
PIER22858EA 195 EUR InStock
PIER22858
Fluorescent Dye Removal Columns, Pierce™
Systèmes de liaison et de marquage des protéines
Pierce™ Dye Removal Columns enable fast and efficient removal of non-reacted fluorescent dyes from protein labeling reactions.
  • Removes free, unconjugated dye from labeled protein solutions in less than a minute
  • Purification resin provides outstanding conjugate recovery
  • Separate resin and microcentrifuge columns allow optimisation for different dyes and concentrations of sample
Removing excess dye after a labeling reaction is often difficult and time-consuming but is essential for accurate determination of dye-to-protein ratios. The dye removal resin in this kit is highly specialised to produce exceptional protein recoveries while effectively removing non-conjugated dye. Using the appropriate amount of resin and buffer conditions, almost any fluorescent dye can be removed with this kit.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité