Quantitative peroxide assay kit (aqueous), Pierce™
Fournisseur: Thermo Fisher Scientific
23280
PIER23280EA 249 EUR InStock
PIER23280
Quantitative peroxide assay kit (aqueous), Pierce™
Tests Tests enzymatiques
Pierce™ Quantitative Peroxide Assay Kit (Aqueous) detects and measures hydrogen peroxide levels in biological samples using an iron and xylenol orange* (XO) reagent.
  • Ready to use – removes the variability, difficulty and expense of formulating in-house
  • Ideal for sensitive measurement of peroxide accumulation in non-lipid samples
This aqueous-compatible peroxide assay kit detects peroxides based on oxidation of ferrous to ferric ion in the presence of xylenol orange. The kit formulation includes sorbitol, which provides sensitivity enhancement. Most proteins do not interfere with this assay, although some metal chelators may require use of a blank (ie. control) to mitigate these effects.
*Xylenol orange is (o-cresolsulfonephthalein-3´-3´-bis-[methyliminodiacetic acid sodium salt]).
Information de livraison: Complete kit includes Reagent A (1 ml) and Reagent B (4×25 ml).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité