Pegylation reagent, MT(PEG)₄ methyl-PEG-thiol compound
Fournisseur: Thermo Fisher Scientific
26132
PIER26132EA 331 EUR InStock
PIER26132
Pegylation reagent, MT(PEG)₄ methyl-PEG-thiol compound
Protéines purification Réactifs de purification des protéines
Methyl-PEG4-thiol is a methyl- and sulfhydryl-terminated compound that contains a 4-unit polyethylene glycol (PEG) spacer and is used to modify surfaces such as quantum dots, monolayers and magnetic particles.
  • PEG groups are flexible, non-immunogenic and hydrophilic
  • Terminal monodentate thiol reacts spontaneously with gold surfaces through dative binding
  • Terminal methyl group coats quantum dot and other surfaces to reduce nonspecific binding in assays
The use of MT(PEG)4 with CT(PEG)12 in surface modification can form a hydrophilic "lawn" of methyl ether-terminated PEGs with periodic exposed carboxylic acid-containing PEGs. The exposed carboxylic acid groups can be coupled to affinity ligands using the carbodiimide coupling reaction with EDC and sulpho-NHS.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité