Crosslink immunoprecipitation (IP) kit, Pierce™
Fournisseur: Thermo Fisher Scientific
26147
PIER26147EA 491 EUR InStock
PIER26147
Crosslink immunoprecipitation (IP) kit, Pierce™
Systèmes de liaison et de marquage des protéines
The Pierce™ Crosslink Immunoprecipitation (IP) Kit enables highly effective and efficient antigen immunoprecipitations by covalently crosslinking antibodies onto Protein A/G resin.
  • Eliminates antibody contamination
  • Immobilise 100 to 200 µg of antibody to prepare ready-made immunoprecipitation affinity resin (reusable) for many experiments
  • Spin columns eliminate resin loss and provide for more efficient separation of solutions
The kit method involves capturing the IP antibody to protein A/G Agarose resin and covalently immobilising it to the support by crosslinking with disuccinmidyl suberate.
Information de livraison: This kit includes protein A/G plus agarose 0,55 ml; DSS crosslinker, 8×2 mg; coupling buffer (20X), 25 ml; IP lysis/wash buffer, 2×50 ml; conditioning buffer (100X), 5 ml; tris-buffered saline (20X), 25 ml; elution buffer, 50 ml; lane marker sample buffer (5X), 5 ml; control agarose resin, 2 ml; spin columns; and collection tubes.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité