Horseradish peroxidase, Pierce™
Fournisseur: Thermo Fisher Scientific
31490
PIER31490EA 113 EUR InStock
PIER31490
Horseradish peroxidase, Pierce™
Réactifs d'électrophorèse Western Blotting Reagents
Pierce™ Horseradish peroxidase (HRP) is purified horseradish peroxidase enzyme for use in activity assays and conjugation to antibodies for ELISA, Western blot and immunohistochemistry applications.
  • Specific activity is greater than 300 units/mg
  • HRP is smaller (40 kDa) than alkaline phosphatase (AP, 140 kDa)
  • Higher specific enzyme activity than both AP and beta-galactosidase (b-Gal)
This purified horseradish peroxidase (HRP) is supplied lyophilised as a salt-free powder for reconstitution and use in protein research methods. The main application for HRP in molecular biology and protein research is as a reporter system for immunoassays and other probe-based assay techniques such as ELISA, Western blotting, EMSA and Southern blotting. The enzyme is usually conjugated to specific secondary antibodies or streptavidin, and its activity is detected with a colour-forming (or light-generating) substrate.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité