Peptide retention standard, Pierce™
Fournisseur: Thermo Fisher Scientific
31700
PIER31700EA 397 EUR InStock
PIER31700
Peptide retention standard, Pierce™
Étalons Étalons organiques
Peptide Retention Standard is a mixture of five c-terminal amide decapeptides that provide reference elution profiles in reverse-phase HPLC.
  • Determine the relative order of peptide elution from a complex mixture
  • Efficiently predict peptide elution profiles to simplify identification and save purification time
  • Identify peptides of known amino acid composition on reverse phase HPLC columns
  • Monitor column performance
  • Evaluate reverse-phase supports of varying n-alkyl chain lengths and ligand densities
The Peptide Retention Standard consists of five C-terminal amide decapeptides, denoted S1 to S5, having the generic formula Ac-Arg-Gly-X-X-Gly-Leu-Gly-Leu-Gly-Lys-Amide. Four of these peptides are Na-acetylated with the sequence variation as follows: Gly3-Gly4, Ala3-Gly4, Val3-Gly4 and Val3-Val4. The fifth peptide, Ala3-Gly4, contains a free Na-amino group. This mixture will provide 100 to 200 injections (approximately 0,1 mg of each peptide) at 0,1 AUFS at 210 nm. The Peptide Retention Standard is supplied as a dry film in a sealed vial.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité