33063
PIER33063EA
217
EUR
InStock
PIER33063
TSAT (Tris-(succinimidyl)aminotriacetate), Pierce™
TSAT (Tris-(succinimidyl)aminotriacetate)
Thermo Scientific Pierce TSAT is tris-(succinimidyl) aminotriacetate, a novel trifunctional crosslinker that contains three short spacers arms linked to a central nitrogen atom, each ending in an amine-reactive NHS-ester group.
- Reactive groups: NHS ester (three ends)
- Reactive towards: amino groups (primary amines)
- NHS ester couples to amines at pH 7-9 form stable amide bonds
- Trifunctional amine-reactive crosslinking agent
- Non-cleavable
- Water-insoluble (dissolve first in DMF or DMSO)
Attention: For Research Use Only. Not for use in diagnostic procedures.