CN/DAB substrate kit, Pierce™
Fournisseur: Thermo Fisher Scientific
34000
PIER34000EA 167 EUR InStock
PIER34000
CN/DAB substrate kit, Pierce™
Réactifs d'électrophorèse Western Blotting Reagents
Pierce™ CN/DAB Substrate Kit is for chromogenic detection of horseradish peroxidase in immunohistochemistry and immunoblotting applications. The formed product is a black precipitate that photographs well and is useful in double staining applications. The coloured product is soluble in xylenes but not soluble in aqueous buffer such as Tris or phosphate. This combination of DAB and CN has a synergistic effect that results in greater sensitivity than either reagent alone.
Information de livraison: Kit contains 4-Chloro-1-naphthol /3,3´-Diaminobenzidine, tetrahydrochloride (10X), 25 ml; Stable Peroxide Substrate Buffer, 250 ml.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité