HRP substrate, 1-Step™ Slow TMB-ELISA
Fournisseur: Thermo Fisher Scientific
34024
PIER34024EA 143 EUR InStock
PIER34024
HRP substrate, 1-Step™ Slow TMB-ELISA
Tests Tests ELISA
The 1-Step™ Slow TMB-ELISA contains soluble TMB (3,3´,5,5´-tetramentylbenzidine) that is ideal for ELISA-based applications. TMB is a chromogenic substrate for horseradish peroxidase. Once oxidised by the enzyme, this substrate yields a blue product that absorbs at 370 nm and 652 nm. Adding sulfuric or phosphoric acid to stop the reaction changes the product to yellow that absorbs at 450 nm and provides greater sensitivity than the blue product. The 1-Step Slow TMB-ELISA is supplied ready to use with no mixing required.
  • No additional reagents or filtering required
  • Non carcinogenic
  • No DMF or DMSO present
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité