SuperSignal™ ELISA Pico Chemiluminescent Substrate
Fournisseur: Thermo Fisher Scientific
37069
PIER37069EA 792 EUR InStock
PIER37069
SuperSignal™ ELISA Pico Chemiluminescent Substrate
Tests Tests ELISA
SuperSignal™ ELISA pico chemiluminescent substrate is optimised to generate an intense light signal and provide exceptional performance in luminometer based assays.
  • Intense signal is produced immediately at room temperature or at 37 °C
  • Signal can be read in black or white opaque microplates
  • Detects protein in ELISAs down to the picogram levels
  • Ambient shipping and room temperature storage
  • Consistent performance of the working solution over an eight hour period
SuperSignal™ ELISA pico chemiluminescent substrate offers greater sensitivity in ELISAs, or any other solution-based assay using chemiluminescent substrates for enzyme detection and quantification. These ELISAs can take place in either a test tube or a microplate and are quantified by measuring relative light units (RLU) in a luminometer.
Information de livraison: Kit contains SuperSignal™ ELISA pico luminol/enhancer solution (50 or 125 ml) and SuperSignal™ ELISA pico luminol/enhancer Solution (50 or 125 ml).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité