Pierce™ Conjugate Purification Kit
Fournisseur: Thermo Fisher Scientific
44920
PIER44920EA 784 EUR InStock
PIER44920
Pierce™ Conjugate Purification Kit
Protéines purification Protein Purification Kits
Pierce™ conjugate purification kit is designed to eliminate excess, unconjugated HRP or alkaline phosphatase enzyme from labeled antibodies (IgG) after performing antibody-enzyme conjugation reactions. The single-use column-kit is intended for purification of 0,5 to 50 mg of antibody conjugate dissolved in tris-buffered saline.
  • Procedure is designed for removal of HRP or AP enzymes from IgG antibodies (not for use with IgM, IgY, Fab fragments or streptavidin conjugates)
  • Removing unconjugated enzyme from antibody conjugates often reduces background and improves overal performance of immunoassays
Information de livraison: Kit includes IDA agarose column, 2 ml (1 column), polyacrylamide desalting column, 10 ml (1 column), Column Activator Buffer (25 ml), Elution Buffer (35 ml), 2× TBS pack (each makes 500 ml), PBS pack (makes 500 ml), and Activity Preservation Solution (1 ml).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité