Fluorescence biotin quantitation kit, Pierce™
Fournisseur: Thermo Fisher Scientific
46610
PIER46610EA 325 EUR InStock
PIER46610
Fluorescence biotin quantitation kit, Pierce™
Systèmes de liaison et de marquage des protéines
Pierce™ Fluorescence Biotin Quantitation Kit requires only 10 μl of sample to accurately measure the biotinylation level of labeled antibodies and other biomolecules.
  • Fast - requires only five minute incubation
  • Economical - requires only 10 µl of sample (~ 750 ng of biotinylated IgG)
  • Sensitive - linear working range of 10 to 60 picomoles of biotin
This microplate-based biotin assay is easy to perform by adding the supplied fluorescent reporter to the biotinylated samples and diluted biocytin standards. The avidin fluoresces when the weakly interacting HABA (4'-hydroxyazobenzene-2-carboxylic acid) is displaced by the biotin. The amount of biotin is determined by comparing the sample's fluorescence to the biocytin standard curve. This assay requires must less sample volume than the microplate colorimetric HABA assay and is much more sensitive.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité