Immunostainer enhancer, Pierce™
Fournisseur: Thermo Fisher Scientific
46644
PIER46644EA 499 EUR InStock
PIER46644
Immunostainer enhancer, Pierce™
Systèmes de liaison et de marquage des protéines
The Pierce™ Immunostain Enhancer is compatible with fluorescent and chromogenic detection and routinely increases both signal intensity and detection sensitivity.
  • Only a fraction of antibody is needed to achieve the same signal as with conventional immunodetection
  • Typically provides 3- to 12-fold increase in signal intensity
  • Sensitivity for improved visualisation of the antigen of interest in cells and tissues
  • Significantly improves signal-to-noise ratio for poor quality and low affinity antibodies
  • Can be used with chromogenic and fluorescent detection methods
It enables significant dilution of primary and secondary antibodies. Dilution 5- to 20-fold beyond the normal has been used successfully; however, the dilution factor varies depending on the antibody, tissue, and antigen.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité