Pierce™ NHS-Fluorescein antibody labeling kit, Pierce™
Fournisseur: Thermo Fisher Scientific
53029
PIER53029EA 426 EUR InStock
PIER53029
Pierce™ NHS-Fluorescein antibody labeling kit, Pierce™
Systèmes de liaison et de marquage des protéines
Pierce™ NHS-Fluorescein antibody labeling kit is a multi-component kit containing a high-performance derivative of fluorescein dye, activated for easy and reliable labeling of antibodies, proteins and other molecules for use as fluorescent probes.
  • Fluorescently label and purify protein in approximately one hour
  • Amine specific labeling - the isothiocyanate variety of fluorescein efficiently labels antibodies and other purified proteins at primary amines
  • Single-use vials of reagent for added convenience
Information de livraison: Standard kit includes NHS Fluorescein (3×50 µg vials), borate buffer (1 ml), spin columns (6 each), collection tubes (12 each) and purification resin (5 ml).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité