Hydrazide resin, UltraLink™
Fournisseur: Thermo Fisher Scientific
53149
PIER53149EA 325 EUR InStock
PIER53149
Hydrazide resin, UltraLink™
Protéines purification Protein Purification Resins
UltraLink™ Hydrazide resin is used to prepare affinity columns with antibodies or other glycolsylated proteins.
  • Able to immobilise 1 to 10 mg of oxidized antibody or other glycoprotein per ml of resin
  • Immobilise in as few as 30 minutes; achieve at least 90% coupling of most glycoproteins in less than 4 hours (efficiency depends on the amount and type of glycosylation)
  • Conjugates only to purified glycoproteins whose sugar groups have been gently oxidised with periodate
  • No stabilisation step is required
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité