Carrier proteins, maleimide-activated carrier proteins, Imject™ Pierce™
Fournisseur: Thermo Fisher Scientific
77605 77606 77610 77663
PIEA77605EA 0 EUR InStock
PIEA77605 PIEA77606 PIER77610 PIEA77663
Carrier proteins, maleimide-activated carrier proteins, Imject™ Pierce™
Protéines extraction Protein Extraction Kits
Imject® Maleimide-activated mcKLH and PEGylated mcKLH enable conjugation of sulfhydryl-containing peptide haptens to elicit an immune response and antibody production against the hapten.
  • Used to achieve the highest antigen conjugation efficiency
  • Cysteine reactive chemistry enables controlled orientation of your antigen
  • High solubility minimises loss during conjugation
The mcKLH is maleimide-activated using a heterobifunctional crosslinker that contains an N-hydroxysuccinimide (NHS) ester and a maleimide group. The PEGylated mcKLH is activated with NHS-PEG₂-Maleimide, creating a more soluble maleimideactivated mcKLH. Maleimide groups react with free sulfhydryls, forming stable thioether bonds.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité