Immobilisation Kit, Glycolink™, Pierce™
Fournisseur: Thermo Fisher Scientific
88941
PIER88941EA 838 EUR InStock
PIER88941
Immobilisation Kit, Glycolink™, Pierce™
Protéines purification Protein Purification Kits
The GlycoLink™ Immobilisation Kit contains affinity columns and reagents to immobilise glycoproteins through oxidised sugar groups for use in affinity-purification applications or studies of protein-protein interactions. This kit is ideal for purification of antigens using covalently coupled antibodies, determining binding partners for a glycoprotein of interest and for performing large-scale affinity purifications.
  • High capacity with 1 to 10 mg of oxidised antibody or other glycoprotein per ml of resin
  • Immobilise in as little as 30 minutes; achieve at least 90% coupling of most glycoproteins in less than 4 hours
  • Hydrazide-activated UltraLink resin conjugates only to purified glycoproteins whose sugar (eg. sialic acid) groups have been gently oxidised with periodate
  • Resonance structure of the hydrazone bonds are sufficiently stable for multiple affinity purifications with one batch of prepared resin; no stabilisation step required
  • Immobilises IgG via the Fc region, thereby keeping both antigen binding sites available for capturing target
Antibodies immobilised by this hydrazide immobilisation chemistry have unobstructed antigen-binding sites and optimal purification capability. These columns can be regenerated and reused five times without loss in binding capacity.
Information de livraison: Kit contains GlycoLink™ columns (1 ml, 10 columns), GlycoLink™ Coupling Buffer (2×100 ml), aniline (0,2 ml), Wash Buffer (120 ml), sodium meta-periodate (0,5 g), Zeba™ spin desalting columns (5 ml, 10 columns), PBS pack (makes 500 ml, 1 pack) and column accessories (10 each).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité