Cobalt spin filter plates, 96 well, HisPur™
Fournisseur: Thermo Fisher Scientific
90095
PIER90095EA 510 EUR InStock
PIER90095
Cobalt spin filter plates, 96 well, HisPur™
Microplaques Plaques de filtration
HisPur™ Cobalt spin plates are convenient, ready-to-use, pre-dispensed filter plates for the efficient purification of polyhistidine-tagged proteins from bacterial, mammalian and baculovirus-infected cells. These spin plates are ideal for simultaneous processing of multiple small (1 mg/well binding capacity) samples.
  • Obtain more than 10 mg of pure His-tagged protein per ml of resin without optimising imidazole washing conditions
  • Cobalt-chelate coordination core binds fewer host protein contaminants, resulting in lower background than nickel resins
  • No metal contamination in eluted histidine-tagged protein sample
  • Purify proteins under native or denaturing conditions
  • Resin can be reused several times or discarded after single use
The spin plates contain a tetradentate chelating agarose resin charged with divalent cobalt (Co2+) for obtaining high-purity his-tagged proteins. This resin binds histidine-tagged proteins with higher specificity (less off-target binding) and releases them with gentler conditions (lower concentrations of imidazole).
Information de livraison: Includes two 96-well filter plates with each well containing 100 µl of resin bed (400 µl of 25% resin slurry), and six wash/collection plates.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité