Protein labelling reagent, iodoTMTzero™
Fournisseur: Thermo Fisher Scientific
90100
PIER90100EA 262 EUR InStock
PIER90100
Protein labelling reagent, iodoTMTzero™
Systèmes de liaison et de marquage des protéines
The iodoTMTzero™ label reagent can be used as an isotopic 'light' duplex tag for MS-level protein quantitation.
  • Only reacts with sulfhydryl groups (reduced, unmodified cysteine residues), thus making it specific
  • Labeled proteins and peptides are not susceptible to reducing agents
  • Options for duplex isotopic (MS) or sixplex isobaric (MS/MS) quantitation
  • Can be used to study cysteine modifications (e.g. di-sulfide bridges, S-nitrosylation) making it versatile
This tag, along with the iodoTMTsixplex™ reagents can enable the quantitation of protein expression changes in cell-based and tissue samples that may not be amenable to metabolic isotopic labeling strategies (eg. SILAC).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité