Rapid equilbrium dialysis (RED) device, Pierce™
Fournisseur: Thermo Fisher Scientific
89811 90006 90007 90085 90087 90088 99006 90112 91012
PIER89811EA 550 EUR InStock
PIER89811 PIER90006 PIER90007 PIER90085 PIER90087 PIER90088 PIER99006 PIER90112 PIER91012
Rapid equilbrium dialysis (RED) device, Pierce™
Tubes Tubes de dialyse
Single-Use RED (rapid equilibrium dialysis) Plate is composed of disposable high-density polypropylene and is pre-loaded with 48 equilibrium dialysis membrane inserts. Each insert is comprised of two side-by-side chambers separated by an O-ring-sealed vertical cylinder of dialysis membrane with varying molecular-weight cutoffs (8 or 12 kD MWCO).
  • Easy and ready to use
  • Design enables equilibrium to reach 200 min⁻¹ agitation with in three to four hours
  • Reproducible and accurate for plasma binding assays
  • Robust compartmentalised design eliminates potential for
More Product Information
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité