Protease Inhibitor Tablets, EDTA-free, contain broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation during cell lysis and protein extraction experiments. 1 * 30 Tablet
Fournisseur: Thermo Fisher Scientific
COMMANDE
Développer tout / Réduire tout
Protease Inhibitor Tablets, EDTA-free, contain broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation during cell lysis and protein extraction experiments. 1 * 30 Tablet PIERA32953 1 * 30 Tablet
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou